Anti-DGKB Rabbit Polyclonal Antibody [clone: 415CT16.3.2.1]

Lieferant: Abgent
ABGEAP8127BEA 360 EUR
ABGEAP8127B
Anti-DGKB Rabbit Polyclonal Antibody [clone: 415CT16.3.2.1]
Antikörper
Type: Primary
Antigen: DGKB (diacylglycerol kinase, beta 90kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR