Anti-GSK3B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8121AEA 360 EUR
ABGEAP8121A
Anti-GSK3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR