Anti-NME2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8081AEA 348 EUR
ABGEAP8081A
Anti-NME2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NME2 (NME/NM23 nucleoside diphosphate kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR