Anti-GNB2L1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8063AEA 350 EUR
ABGEAP8063A
Anti-GNB2L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GNB2L1 (guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR