Anti-PIP5K1C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8039BEA 360 EUR
ABGEAP8039B
Anti-PIP5K1C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIP5K1C (phosphatidylinositol-4-phosphate 5-kinase, type I, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR