Anti-PI3KCA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8016BEA 360 EUR
ABGEAP8016B
Anti-PI3KCA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PI3KCA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR