Anti-CYP3A43 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7999CEA 360 EUR
ABGEAP7999C
Anti-CYP3A43 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP3A43 (cytochrome P450, family 3, subfamily A, polypeptide 43)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR