Anti-FGF10 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7975BEA 360 EUR
ABGEAP7975B
Anti-FGF10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGF10 (fibroblast growth factor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR