Anti-CASP9 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7974CEA 348 EUR
ABGEAP7974C
Anti-CASP9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CASP9 (caspase 9, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR