Anti-MAP3K14 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7967CEA 348 EUR
ABGEAP7967C
Anti-MAP3K14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR