Anti-PRKX Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7940AEA 360 EUR
ABGEAP7940A
Anti-PRKX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRKX (protein kinase, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR