Anti-PAK4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7929AEA 350 EUR
ABGEAP7929A
Anti-PAK4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAK4 (p21 protein (Cdc42/Rac)-activated kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR