Anti-PAK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7927AEA 348 EUR
ABGEAP7927A
Anti-PAK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAK2 (p21 protein (Cdc42/Rac)-activated kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR