Anti-MAP2K5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7906FEA 350 EUR
ABGEAP7906F
Anti-MAP2K5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP2K5 (mitogen-activated protein kinase kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR