Anti-CYP4F12 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7895BEA 348 EUR
ABGEAP7895B
Anti-CYP4F12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP4F12 (cytochrome P450, family 4, subfamily F, polypeptide 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR