Anti-CYP4A11 (4A22) Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7884BEA 350 EUR
ABGEAP7884B
Anti-CYP4A11 (4A22) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP4A11 (4A22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR