Anti-CYP21A2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7880AEA 350 EUR
ABGEAP7880A
Anti-CYP21A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP21A2 (cytochrome P450, family 21, subfamily A, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR