Anti-GAPDH Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7873AEA 370 EUR
ABGEAP7873A
Anti-GAPDH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR