Anti-MERTK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7865AEA 348 EUR
ABGEAP7865A
Anti-MERTK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MERTK (c-mer proto-oncogene tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR