Anti-ALDH3A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7849AEA 350 EUR
ABGEAP7849A
Anti-ALDH3A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH3A1 (aldehyde dehydrogenase 3 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR