Anti-CARS2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7846BEA 350 EUR
ABGEAP7846B
Anti-CARS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CARS2 (cysteinyl-tRNA synthetase 2, mitochondrial (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR