Anti-ARAF Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7809CEA 348 EUR
ABGEAP7809C
Anti-ARAF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARAF (v-raf murine sarcoma 3611 viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR