Anti-ACVR1B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7801CEA 350 EUR
ABGEAP7801C
Anti-ACVR1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACVR1B (activin A receptor, type IB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR