Anti-ACTN4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7790BEA 348 EUR
ABGEAP7790B
Anti-ACTN4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACTN4 (actinin, alpha 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR