Anti-LYN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7713AEA 348 EUR
ABGEAP7713A
Anti-LYN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR