Anti-ABL1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7694CEA 348 EUR
ABGEAP7694C
Anti-ABL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ABL1 (c-abl oncogene 1, non-receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR