Anti-RYK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7677AEA 360 EUR
ABGEAP7677A
Anti-RYK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RYK (receptor-like tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR