Anti-MST1R Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7674CEA 348 EUR
ABGEAP7674C
Anti-MST1R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MST1R (macrophage stimulating 1 receptor (c-met-related tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR