Anti-ROR2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7672AEA 360 EUR
ABGEAP7672A
Anti-ROR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ROR2 (receptor tyrosine kinase-like orphan receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR