Anti-IGF1R Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7649AEA 348 EUR
ABGEAP7649A
Anti-IGF1R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR