Anti-FGFR2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7637AEA 348 EUR
ABGEAP7637A
Anti-FGFR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR