Anti-AXL Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7602DEA 350 EUR
ABGEAP7602D
Anti-AXL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AXL (AXL receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR