Anti-PTPN7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7599AEA 348 EUR
ABGEAP7599A
Anti-PTPN7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPN7 (protein tyrosine phosphatase, non-receptor type 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR