Anti-MAPK8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7505AEA 360 EUR
ABGEAP7505A
Anti-MAPK8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPK8 (mitogen-activated protein kinase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR