Anti-EN2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7457FEA 350 EUR
ABGEAP7457F
Anti-EN2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EN2 (engrailed homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR