Anti-FKBP9 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7388BEA 350 EUR
ABGEAP7388B
Anti-FKBP9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FKBP9 (FK506 binding protein 9, 63 kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR