Anti-FKBP4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7387CEA 348 EUR
ABGEAP7387C
Anti-FKBP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FKBP4 (FK506 binding protein 4, 59kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR