Anti-S100A6 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7346BEA 350 EUR
ABGEAP7346B
Anti-S100A6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S100A6 (S100 calcium binding protein A6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR