Anti-TYR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7345CEA 350 EUR
ABGEAP7345C
Anti-TYR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TYR (tyrosinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR