Anti-KLF5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7342CEA 350 EUR
ABGEAP7342C
Anti-KLF5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLF5 (Kruppel-like factor 5 (intestinal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR