Anti-POLE3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7316AEA 360 EUR
ABGEAP7316A
Anti-POLE3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POLE3 (polymerase (DNA directed), epsilon 3, accessory subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR