Anti-HOMER1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7302AEA 348 EUR
ABGEAP7302A
Anti-HOMER1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOMER1 (homer homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR