Anti-NAE1 (APPBP1) Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7273EEA 350 EUR
ABGEAP7273E
Anti-NAE1 (APPBP1) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAE1 (APPBP1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR