Anti-ADRB2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7263FEA 348 EUR
ABGEAP7263F
Anti-ADRB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADRB2 (adrenoceptor beta 2, surface)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR