Anti-PHKG2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7233AEA 350 EUR
ABGEAP7233A
Anti-PHKG2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PHKG2 (phosphorylase kinase, gamma 2 (testis))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR