Anti-MAPK14 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7226FEA 350 EUR
ABGEAP7226F
Anti-MAPK14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPK14 (mitogen-activated protein kinase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR