Anti-PRAK (MAPKAPK5) Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7216AEA 350 EUR
ABGEAP7216A
Anti-PRAK (MAPKAPK5) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRAK (MAPKAPK5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR