Anti-CASK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7212AEA 348 EUR
ABGEAP7212A
Anti-CASK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CASK (calcium/calmodulin-dependent serine protein kinase (MAGUK family))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR