Anti-CAMK1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7205AEA 348 EUR
ABGEAP7205A
Anti-CAMK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMK1 (calcium/calmodulin-dependent protein kinase I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR