Anti-AKR7A2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7186AEA 348 EUR
ABGEAP7186A
Anti-AKR7A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR7A2 (aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR